2-Bromo-5-(trifluoromethyl)pyridin-3-amine 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC910102-100MG
PC910102-250MG
APOSPC910102-100MGEA
45.1
EUR
APOSPC910102-100MG
APOSPC910102-250MG
2-Bromo-5-(trifluoromethyl)pyridin-3-amine 98%
2-Bromo-5-(trifluoromethyl)pyridin-3-amine
Formel:
C₆H₄BrF₃N₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD14702313 CAS-Nummer: 1211515-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...