7-Fluorobenzo[b]thiophene 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC910028-1G
PC910028-250MG
APOSPC910028-1GEA
132
EUR
APOSPC910028-1G
APOSPC910028-250MG
7-Fluorobenzo[b]thiophene 95%
7-Fluorobenzo[b]thiophene
Formel:
C₈H₅FS Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD17169311 CAS-Nummer: 346592-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...