1-Ethoxy-3-fluoro-2-nitrobenzene 95%

Lieferant: Apollo Scientific

PC909882-1G PC909882-250MG PC909882-5G
APOSPC909882-1GEA 230 EUR
APOSPC909882-1G APOSPC909882-250MG APOSPC909882-5G
1-Ethoxy-3-fluoro-2-nitrobenzene 95%
1-Ethoxy-3-fluoro-2-nitrobenzene
Formel: C₈H₈FNO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13689265
CAS-Nummer: 1233952-97-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR