3-Fluoro-5-phenylpyridine 97%

Lieferant: Apollo Scientific

PC909608-1G PC909608-250MG
APOSPC909608-1GEA 632 EUR
APOSPC909608-1G APOSPC909608-250MG
3-Fluoro-5-phenylpyridine 97%
3-Fluoro-5-phenylpyridine
Formel: C₁₁H₈FN
Molekulargewicht: 173.186 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD14701970
CAS-Nummer: 1214374-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR