N-Ethyl-5-fluoro-2-nitroaniline 97%

Lieferant: Apollo Scientific

PC909139-1G PC909139-5G
APOSPC909139-1GEA 219 EUR
APOSPC909139-1G APOSPC909139-5G
N-Ethyl-5-fluoro-2-nitroaniline 97%
N-Ethyl-5-fluoro-2-nitroaniline
Formel: C₈H₉FN₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16744215
CAS-Nummer: 475278-46-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR