2,4,5-Trifluoro-3-ethoxybenzoic acid 95%
Lieferant: Apollo Scientific
PC907097-1G
PC907097-5G
APOSPC907097-1GEA
309
EUR
APOSPC907097-1G
APOSPC907097-5G
2,4,5-Trifluoro-3-ethoxybenzoic acid 95%
2,4,5-Trifluoro-3-ethoxybenzoic acid
Formel:
C₉H₇F₃O₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08459286 CAS-Nummer: 169507-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...