4,4-Difluorocyclohexaneethanamine 95%

Lieferant: Apollo Scientific

PC904995-1G PC904995-250MG
APOSPC904995-1GEA 474 EUR
APOSPC904995-1G APOSPC904995-250MG
4,4-Difluorocyclohexaneethanamine 95%
4,4-Difluorocyclohexaneethanamine
Formel: C₈H₁₅F₂N
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13189567
CAS-Nummer: 1054314-53-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR