3-Ethenyl-5-fluoropyridine 95%

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-5-vinylpyridine

PC904618-1G PC904618-250MG
APOSPC904618-1GEA 243 EUR
APOSPC904618-1G APOSPC904618-250MG
3-Ethenyl-5-fluoropyridine 95%
3-Ethenyl-5-fluoropyridine
Formel: C₇H₆FN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18384200
CAS-Nummer: 1133879-69-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR