Ethyl 2-fluoro-6-methylbenzoate 97%
Lieferant: Apollo Scientific
PC903790-10G
PC903790-1G
PC903790-5G
APOSPC903790-10GEA
754
EUR
APOSPC903790-10G
APOSPC903790-1G
APOSPC903790-5G
Ethyl 2-fluoro-6-methylbenzoate 97%
Ethyl 2-fluoro-6-methylbenzoate
Formel:
C₁₀H₁₁FO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11870183 CAS-Nummer: 90259-30-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...