3-Bromo-5-chloro-2-fluorophenol 95%

Lieferant: Apollo Scientific

PC903363-1G PC903363-250MG PC903363-5G PC903363-10G
APOSPC903363-1GEA 328 EUR
APOSPC903363-1G APOSPC903363-250MG APOSPC903363-5G APOSPC903363-10G
3-Bromo-5-chloro-2-fluorophenol 95%
3-Bromo-5-chloro-2-fluorophenol
Formel: C₆H₃BrClFO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28143932
CAS-Nummer: 1804897-55-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR