6,7-Difluoro-1-methyl-1,3-benzimidazole 98%
Lieferant: Apollo Scientific
PC902264-1G
PC902264-5G
APOSPC902264-1GEA
219
EUR
APOSPC902264-1G
APOSPC902264-5G
6,7-Difluoro-1-methyl-1,3-benzimidazole 98%
6,7-Difluoro-1-methyl-1,3-benzimidazole
Formel:
C₈H₆F₂N₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD20529460 CAS-Nummer: 1352318-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...