6,7-Difluoro-1-isopropylbenzoimidazole 97%

Lieferant: Apollo Scientific

PC902259-1G PC902259-5G PC902259-25G
APOSPC902259-1GEA 132 EUR
APOSPC902259-1G APOSPC902259-5G APOSPC902259-25G
6,7-Difluoro-1-isopropylbenzoimidazole 97%
6,7-Difluoro-1-isopropylbenzoimidazole
Formel: C₁₀H₁₀F₂N₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD19981547
CAS-Nummer: 1330750-46-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR