1-Propyl-3-(trifluoromethyl)pyrazole 95%
Lieferant: Apollo Scientific
PC902062-1G
PC902062-5G
APOSPC902062-1GEA
219
EUR
APOSPC902062-1G
APOSPC902062-5G
1-Propyl-3-(trifluoromethyl)pyrazole 95%
1-Propyl-3-(trifluoromethyl)pyrazole
Formel:
C₇H₉F₃N₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16619915 CAS-Nummer: 1033132-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...