2-(Cyclopropylmethoxy)-5-fluoropyridine-4-boronic acid

Lieferant: Apollo Scientific
Achtung

PC901405-1G PC901405-5G
APOSPC901405-1GEA 306 EUR
APOSPC901405-1G APOSPC901405-5G
2-(Cyclopropylmethoxy)-5-fluoropyridine-4-boronic acid
2-(Cyclopropylmethoxy)-5-fluoropyridine-4-boronic acid
Formel: C₉H₁₁BFNO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18914506
CAS-Nummer: 2096334-21-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR