3-Ethoxy-2-fluoro-5-methylphenylboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (3-Ethoxy-2-fluoro-5-methylphenyl)boronic acid

PC901404-1G PC901404-5G
APOSPC901404-1GEA 219 EUR
APOSPC901404-1G APOSPC901404-5G
3-Ethoxy-2-fluoro-5-methylphenylboronic acid 98%
3-Ethoxy-2-fluoro-5-methylphenylboronic acid
Formel: C₉H₁₂BFO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD19981525
CAS-Nummer: 2096333-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR