2-Chloro-5-fluoro-4-methylphenylboronic acid 98%

Lieferant: Apollo Scientific

PC900998-1G PC900998-5G
APOSPC900998-1GEA 219 EUR
APOSPC900998-1G APOSPC900998-5G
2-Chloro-5-fluoro-4-methylphenylboronic acid 98%
2-Chloro-5-fluoro-4-methylphenylboronic acid
Formel: C₇H₇BClFO₂
Molekulargewicht: 188.392 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20441814
CAS-Nummer: 1612184-35-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR