2-(Aminomethyl)-3,5-difluoropyridine dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (3,5-difluoropyridin-2-yl)methanamine dihydrochloride

PC8929-1G PC8929-250MG
APOSPC8929-1GEA 259 EUR
APOSPC8929-1G APOSPC8929-250MG
2-(Aminomethyl)-3,5-difluoropyridine dihydrochloride
2-(Aminomethyl)-3,5-difluoropyridine dihydrochloride
Formel: C₆H₆F₂N₂·2ClH
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD12827699
CAS-Nummer: 1204298-48-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR