1-[8-Fluoro-2-(trifluoromethyl)quinolin-4-yl]piperazine 97%

Lieferant: Apollo Scientific

PC8870-1G PC8870-500MG
APOSPC8870-1GEA 263 EUR
APOSPC8870-1G APOSPC8870-500MG
1-[8-Fluoro-2-(trifluoromethyl)quinolin-4-yl]piperazine 97%
1-[8-Fluoro-2-(trifluoromethyl)quinolin-4-yl]piperazine
Formel: C₁₄H₁₃F₄N₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03094528
CAS-Nummer: 401567-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR