2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzylamine 97%
Lieferant: Apollo Scientific
PC8771-1G
PC8771-250MG
APOSPC8771-1GEA
230
EUR
APOSPC8771-1G
APOSPC8771-250MG
2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzylamine 97%
2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzylamine
Formel:
C₁₂H₁₂F₃N₃ Siedepunkt: 120 °C (0.1 mm Hg) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09817513 CAS-Nummer: 910095-35-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...