1-Isopropyl-2-(trifluoromethyl)-1H-benzimidazole-5-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Prop-2-yl)-2-(trifluoromethyl)-1H-benzimidazole-5-carboxylic acid, 5-Carboxy-1-isopropyl-2-(trifluoromethyl)-1H-benzimidazole

PC8566-1G PC8566-10G PC8566-5G
APOSPC8566-1GEA 23.2 EUR
APOSPC8566-1G APOSPC8566-10G APOSPC8566-5G
1-Isopropyl-2-(trifluoromethyl)-1H-benzimidazole-5-carboxylic acid 98%
1-Isopropyl-2-(trifluoromethyl)-1H-benzimidazole-5-carboxylic acid
Formel: C₁₂H₁₁F₃N₂O₂
Schmelzpunkt: 174…178 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01570675
CAS-Nummer: 306935-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR