4-Bromo-2,3,6-trifluorobenzyl bromide 95%
Lieferant: Apollo Scientific
PC8249-1G
PC8249-250MG
APOSPC8249-1GEA
632
EUR
APOSPC8249-1G
APOSPC8249-250MG
4-Bromo-2,3,6-trifluorobenzyl bromide 95%
4-Bromo-2,3,6-trifluorobenzyl bromide
Formel:
C₇H₃Br₂F₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03094470 CAS-Nummer: 537033-57-1 UN-Nummer: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...