5-Bromo-4-chloro-2-fluorobenzoic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC7755-250MG
PC7755-1G
PC7755-5G
APOSPC7755-250MGEA
21.9
EUR
APOSPC7755-250MG
APOSPC7755-1G
APOSPC7755-5G
5-Bromo-4-chloro-2-fluorobenzoic acid 98%
5-Bromo-4-chloro-2-fluorobenzoic acid
Formel:
C₇H₃BrClFO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00672928 CAS-Nummer: 289038-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...