3,5-Difluoroisonicotinaldehyde 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Difluoro-4-formylpyridine, 3,5-Difluoropyridine-4-carboxaldehyde
PC7022-1G
PC7022-5G
APOSPC7022-1GEA
285
EUR
APOSPC7022-1G
APOSPC7022-5G
3,5-Difluoroisonicotinaldehyde 98%
3,5-Difluoroisonicotinaldehyde
Formel:
C₆H₃F₂NO Schmelzpunkt: 51…52 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07368235 CAS-Nummer: 870234-98-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...