2,3-Difluoro-4-(trifluoromethyl)benzeneboronic acid 96+%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,3-Difluoro-4-(trifluoromethyl)phenyl)boronic acid, [2,3-Difluoro-4-(trifluoromethyl)phenyl]boronic acid

PC6952-5G PC6952-1G PC6952-25G
APOSPC6952-5GEA 438 EUR
APOSPC6952-5G APOSPC6952-1G APOSPC6952-25G
2,3-Difluoro-4-(trifluoromethyl)benzeneboronic acid 96+%
2,3-Difluoro-4-(trifluoromethyl)benzeneboronic acid
Formel: C₇H₄BF₅O₂
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD14892600
CAS-Nummer: 2096332-61-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR