4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pent-2-enoic acid 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-(Perfluoroisopropyl)acrylic acid, 3-(Heptafluoroprop-2-yl)prop-2-enoic acid

PC6932L-1G PC6932L-5G PC6932L-25G
APOSPC6932L-1GEA 36.4 EUR
APOSPC6932L-1G APOSPC6932L-5G APOSPC6932L-25G
4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pent-2-enoic acid 95%
4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pent-2-enoic acid
Formel: C₆H₃F₇O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09998187
CAS-Nummer: 243139-64-2
UN-Nummer: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR