3,5-Difluoro-DL-phenylglycine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Amino(3,5-difluorophenyl)acetic acid, 2-Amino-2-(3,5-difluorophenyl)ethanoic acid
PC6579-1G
PC6579-5G
APOSPC6579-1GEA
116
EUR
APOSPC6579-1G
APOSPC6579-5G
3,5-Difluoro-DL-phenylglycine 98%
3,5-Difluoro-DL-phenylglycine
Formel:
C₈H₇F₂NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06660157 CAS-Nummer: 199327-33-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...