2-Fluoro-3-(trifluoromethoxy)benzyl alcohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[2-Fluoro-3-(trifluoromethoxy)phenyl]methanol
PC6311-1G
PC6311-5G
APOSPC6311-1GEA
69.6
EUR
APOSPC6311-1G
APOSPC6311-5G
2-Fluoro-3-(trifluoromethoxy)benzyl alcohol
2-Fluoro-3-(trifluoromethoxy)benzyl alcohol
Formel:
C₈H₆F₄O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12026473 CAS-Nummer: 86256-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...