4-(Methylsulphonyl)-3-(trifluoromethyl)benzonitrile 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Cyano-2-(methylsulphonyl)benzotrifluoride, 4-Cyano-2-(trifluoromethyl)phenyl methyl sulphone
PC6219-1G
PC6219-5G
APOSPC6219-1GEA
415
EUR
APOSPC6219-1G
APOSPC6219-5G
4-(Methylsulphonyl)-3-(trifluoromethyl)benzonitrile 98%
4-(Methylsulphonyl)-3-(trifluoromethyl)benzonitrile
Formel:
C₉H₆F₃NO₂S Schmelzpunkt: 171…172 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12196930 CAS-Nummer: 904311-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...