2-Chloro-1,1,1,6,6,6-hexafluorohexa-2,4-diene

Lieferant: Apollo Scientific
Achtung

PC5778-1G PC5778-5G
APOSPC5778-1GEA 157 EUR
APOSPC5778-1G APOSPC5778-5G
2-Chloro-1,1,1,6,6,6-hexafluorohexa-2,4-diene
2-Chloro-1,1,1,6,6,6-hexafluorohexa-2,4-diene
Formel: C₆H₃ClF₆
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08461616
CAS-Nummer: 885276-10-8
UN-Nummer: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR