2-Bromo-6-chloro-4-(trifluoromethyl)benzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

PC56968-1G PC56968-250MG PC56968-5G
APOSPC56968-1GEA 665 EUR
APOSPC56968-1G APOSPC56968-250MG APOSPC56968-5G
2-Bromo-6-chloro-4-(trifluoromethyl)benzaldehyde 95%
2-Bromo-6-chloro-4-(trifluoromethyl)benzaldehyde
Formel: C₈H₃BrClF₃O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD31812115
CAS-Nummer: 1415130-43-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR