2-Chloro-4-(trifluoromethoxy)benzyl alcohol 95%

Lieferant: Apollo Scientific

Synonyme: (2-Chloro-4-(trifluoromethoxy)phenyl)methanol

PC56777-1G PC56777-5G
APOSPC56777-1GEA 473 EUR
APOSPC56777-1G APOSPC56777-5G
2-Chloro-4-(trifluoromethoxy)benzyl alcohol 95%
2-Chloro-4-(trifluoromethoxy)benzyl alcohol
Formel: C₈H₆ClF₃O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18393815
CAS-Nummer: 1261822-74-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR