2-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid 98%

Lieferant: Apollo Scientific
Achtung

PC56714-1G PC56714-250MG PC56714-5G
APOSPC56714-1GEA 290 EUR
APOSPC56714-1G APOSPC56714-250MG APOSPC56714-5G
2-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid 98%
2-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid
Formel: C₈H₃F₄NO₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22273734
CAS-Nummer: 1389313-52-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR