2-Bromo-3-(bromomethyl)-4-fluoro-1-(trifluoromethyl)benzene 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-3-(bromomethyl)-4-fluorobenzotrifluoride, 2-Bromo-6-fluoro-3-(trifluoromethyl)benzyl bromide
PC56252-1G
PC56252-250MG
PC56252-5G
APOSPC56252-1GEA
58
EUR
APOSPC56252-1G
APOSPC56252-250MG
APOSPC56252-5G
2-Bromo-3-(bromomethyl)-4-fluoro-1-(trifluoromethyl)benzene 95%
2-Bromo-3-(bromomethyl)-4-fluoro-1-(trifluoromethyl)benzene
Formel:
C₈H₄Br₂F₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD31580165 CAS-Nummer: 2090283-00-2 UN-Nummer: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...