2-Bromo-6-fluoro-3-(trifluoromethyl)benzyl alcohol 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: [2-Bromo-6-fluoro-3-(trifluoromethyl)phenyl]methanol

PC56211-1G PC56211-250MG
APOSPC56211-1GEA 642 EUR
APOSPC56211-1G APOSPC56211-250MG
2-Bromo-6-fluoro-3-(trifluoromethyl)benzyl alcohol 95+%
2-Bromo-6-fluoro-3-(trifluoromethyl)benzyl alcohol
Formel: C₈H₅BrF₄O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD31580162
CAS-Nummer: 2090690-37-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR