2-Hydroxy-5-iodo-3-(trifluoromethyl)pyridine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Iodo-3-(trifluoromethyl)pyridin-2-ol

PC56189-1G PC56189-5G PC56189-25G
APOSPC56189-1GEA 23.2 EUR
APOSPC56189-1G APOSPC56189-5G APOSPC56189-25G
2-Hydroxy-5-iodo-3-(trifluoromethyl)pyridine 95%
2-Hydroxy-5-iodo-3-(trifluoromethyl)pyridine
Formel: C₆H₃F₃INO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12024303
CAS-Nummer: 887707-23-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR