Potassium 2,2,2-trifluoroethane-2-trifluoroborate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Potassium, trifluoro(2,2,2-trifluoroethyl)boranuide
PC56181-100MG
PC56181-250MG
APOSPC56181-100MGEA
277
EUR
APOSPC56181-100MG
APOSPC56181-250MG
Potassium 2,2,2-trifluoroethane-2-trifluoroborate
Potassium 2,2,2-trifluoroethane-2-trifluoroborate
Formel:
C₂H₂BF₆·K Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD27981361 CAS-Nummer: 1510778-85-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...