2,2,2,2',3'-Pentafluoro-4'-(trifluoromethyl)acetophenone 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[2,3-Difluoro-4-(trifluoromethyl)phenyl]-2,2,2-trifluoroethanone
PC56176-1G
PC56176-250MG
PC56176-5G
APOSPC56176-1GEA
1220
EUR
APOSPC56176-1G
APOSPC56176-250MG
APOSPC56176-5G
2,2,2,2',3'-Pentafluoro-4'-(trifluoromethyl)acetophenone 95+%
2,2,2,2',3'-Pentafluoro-4'-(trifluoromethyl)acetophenone
Formel:
C₉H₂F₈O Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD31568567 CAS-Nummer: 2227272-56-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...