2-Bromo-4-fluoro-3-iodobenzotrifluoride 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-1-fluoro-2-iodo-4-(trifluoromethyl)benzene
PC56173-1G
PC56173-250MG
APOSPC56173-1GEA
1210
EUR
APOSPC56173-1G
APOSPC56173-250MG
2-Bromo-4-fluoro-3-iodobenzotrifluoride 97%
2-Bromo-4-fluoro-3-iodobenzotrifluoride
Formel:
C₇H₂BrF₄I Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD31568566 CAS-Nummer: 2227272-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...