3-(Trifluoromethylsulphonyl)acetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Trifluoromethanesulphonylphenyl)ethanone

PC56111-1G PC56111-250MG
APOSPC56111-1GEA 487 EUR
APOSPC56111-1G APOSPC56111-250MG
3-(Trifluoromethylsulphonyl)acetophenone
3-(Trifluoromethylsulphonyl)acetophenone
Formel: C₉H₇F₃O₃S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD25460316
CAS-Nummer: 1100768-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR