4-Bromo-2-(2,2,2-trifluoroethoxy)pyridine

Lieferant: Apollo Scientific
Achtung

PC56063-1G PC56063-250MG PC56063-5G
APOSPC56063-1GEA 272 EUR
APOSPC56063-1G APOSPC56063-250MG APOSPC56063-5G
4-Bromo-2-(2,2,2-trifluoroethoxy)pyridine
4-Bromo-2-(2,2,2-trifluoroethoxy)pyridine
Formel: C₇H₅BrF₃NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13190652
CAS-Nummer: 161952-62-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR