Methyl 4-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]benzoate 95%

Lieferant: Apollo Scientific

PC53607-1G PC53607-250MG
APOSPC53607-1GEA 574 EUR
APOSPC53607-1G APOSPC53607-250MG
Methyl 4-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]benzoate 95%
Methyl 4-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]benzoate
Formel: C₁₁H₈F₆O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD32663533
CAS-Nummer: 721961-57-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR