Methyl 2,6-dichloro-3-fluoro-benzoate 95%

Lieferant: Apollo Scientific

PC53596-1G PC53596-250MG PC53596-5G
APOSPC53596-1GEA 575 EUR
APOSPC53596-1G APOSPC53596-250MG APOSPC53596-5G
Methyl 2,6-dichloro-3-fluoro-benzoate 95%
Methyl 2,6-dichloro-3-fluoro-benzoate
Formel: C₈H₅Cl₂FO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12406864
CAS-Nummer: 1214353-49-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR