Methyl 5-amino-4-bromo-2-fluorobenzoate
Lieferant: Apollo Scientific
|
Achtung
|
PC53566-1G
PC53566-250MG
APOSPC53566-1GEA
168
EUR
APOSPC53566-1G
APOSPC53566-250MG
Methyl 5-amino-4-bromo-2-fluorobenzoate
Methyl 5-amino-4-bromo-2-fluorobenzoate
Formel:
C₈H₇BrFNO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD17168715 CAS-Nummer: 1342446-00-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...