2,2,3,3-Tetrafluoro-1,4-benzodioxine-6-boronic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC53559-1G
PC53559-250MG
APOSPC53559-1GEA
862
EUR
APOSPC53559-1G
APOSPC53559-250MG
2,2,3,3-Tetrafluoro-1,4-benzodioxine-6-boronic acid
2,2,3,3-Tetrafluoro-1,4-benzodioxine-6-boronic acid
Formel:
C₈H₅BF₄O₄ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD30729940 CAS-Nummer: 622383-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...