Potassium 4-fluorobenzyl-trifluoroborate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Potassium trifluoro(4-fluorobenzyl)borate, Potassium Trifluoro[(4-fluorophenyl)methyl]boranuide
PC53536-1G
PC53536-5G
APOSPC53536-1GEA
84.1
EUR
APOSPC53536-1G
APOSPC53536-5G
Potassium 4-fluorobenzyl-trifluoroborate
Potassium 4-fluorobenzyl-trifluoroborate
Formel:
C₇H₆BF₄·K Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04112715 CAS-Nummer: 1494466-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...