3-(Trifluoromethyl)isonicotinaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC535056-250MG
PC535056-100MG
APOSPC535056-250MGEA
255
EUR
APOSPC535056-250MG
APOSPC535056-100MG
3-(Trifluoromethyl)isonicotinaldehyde 95%
3-(Trifluoromethyl)isonicotinaldehyde
Formel:
C₇H₄F₃NO Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09834785 CAS-Nummer: 1060801-92-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...