2-Fluoro-3-iodoaniline

Lieferant: Apollo Scientific
Achtung

PC53504-1G PC53504-250MG
APOSPC53504-1GEA 719 EUR
APOSPC53504-1G APOSPC53504-250MG
2-Fluoro-3-iodoaniline
2-Fluoro-3-iodoaniline
Formel: C₆H₅FIN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18396913
CAS-Nummer: 1261500-65-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR