5-Bromo-2-methyl-3-(trifluoromethyl)pyridine 97+%
Lieferant: Apollo Scientific
|
Achtung
|
PC535036-1G
PC535036-250MG
PC535036-5G
PC535036-10G
APOSPC535036-1GEA
36.4
EUR
APOSPC535036-1G
APOSPC535036-250MG
APOSPC535036-5G
APOSPC535036-10G
5-Bromo-2-methyl-3-(trifluoromethyl)pyridine 97+%
5-Bromo-2-methyl-3-(trifluoromethyl)pyridine
Formel:
C₇H₅BrF₃N Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD18257687 CAS-Nummer: 1211526-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...