Ethyl 2-amino-5-fluorobenzoate 97%

Lieferant: Apollo Scientific
Achtung

PC535014-1G PC535014-250MG PC535014-5G
APOSPC535014-1GEA 112 EUR
APOSPC535014-1G APOSPC535014-250MG APOSPC535014-5G
Ethyl 2-amino-5-fluorobenzoate 97%
Ethyl 2-amino-5-fluorobenzoate
Formel: C₉H₁₀FNO₂
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD11042774
CAS-Nummer: 391-93-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR