5-Bromo-2-(trifluoromethyl)pyridin-3-amine 97%
Lieferant: Apollo Scientific
PC53491-1G
PC53491-250MG
PC53491-5G
APOSPC53491-1GEA
763
EUR
APOSPC53491-1G
APOSPC53491-250MG
APOSPC53491-5G
5-Bromo-2-(trifluoromethyl)pyridin-3-amine 97%
5-Bromo-2-(trifluoromethyl)pyridin-3-amine
Formel:
C₆H₄BrF₃N₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD18254484 CAS-Nummer: 1211533-18-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...